siRNA Details
Gene Symbol: Bfar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013125.1 | NP_001013143.1 | 4 | 679 | ||
GenBank mRNA | BC087103.1 | - | 4 | 679 | 7192877 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Bfar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013125.1 | NP_001013143.1 | 4 | 679 | ||
GenBank mRNA | BC087103.1 | - | 4 | 679 | 7192877 |