siRNA Details
Gene Symbol: Ccnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107171.1 | NP_001100641.1 | 9 | 1381 | ||
XM_006249686.2 | XP_006249748.1 | 9 | 1389 | |||
XM_006249687.2 | XP_006249749.1 | 5 | 4090 | |||
XM_008769457.1 | XP_008767679.1 | 8 | 1260 | |||
XM_008769458.1 | XP_008767680.1 | 6 | 3166 |
