siRNA Details
Gene Symbol: R3hdm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134867.1 | NP_001128339.1 | 9, 10 | 1141 | ||
XM_006249690.2 | XP_006249752.1 | 9, 10 | 1501 | |||
XM_006249694.2 | XP_006249756.1 | 7, 8 | 1006 | |||
XM_008769459.1 | XP_008767681.1 | 9, 10 | 1138 | |||
XM_008769460.1 | XP_008767682.1 | 9, 10 | 1138 | |||
XM_008769461.1 | XP_008767683.1 | 9, 10 | 1138 | |||
XM_008769462.1 | XP_008767684.1 | 9, 10 | 1138 | |||
XM_008769463.1 | XP_008767685.1 | 7, 8 | 1005 | |||
GenBank mRNA | BC087715.1 | - | 8, 9 | 819 |
