siRNA Details
Gene Symbol: Slc26a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107172.1 | NP_001100642.1 | 16 | 1868 | ||
XM_008769466.1 | XP_008767688.1 | 16 | 2036 |

More Information
Other Species Targeted

Gene Ontology Categories:
