siRNA Details
Gene Symbol: Slc45a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135868.1 | NP_001129340.1 | 3 | 1031 | ||
XM_006249765.2 | XP_006249827.1 | 3 | 1006 | |||
XM_006249766.2 | XP_006249828.1 | 3 | 1131 | |||
XM_006249767.2 | XP_006249829.1 | 3 | 1609 | |||
XM_006249769.2 | XP_006249831.1 | 3 | 1781 |
