siRNA Details
Gene Symbol: Ppp1r15b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107175.1 | NP_001100645.1 | 2 | 2382 | ||
XM_006249781.2 | XP_006249843.1 | 2 | 1454 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_133819, AK170862, AK146089, AK082957, AK049028, and others.
Gene Ontology Categories:
