siRNA Details
Gene Symbol: Trmt1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037192.1 | NP_001032269.1 | 6 | 952 | ||
XM_006249992.2 | XP_006250054.1 | 6 | 1050 | |||
XM_006249993.2 | XP_006250055.1 | 6 | 1055 | |||
XM_008769627.1 | XP_008767849.1 | 7 | 1396 | |||
XM_008769628.1 | XP_008767850.1 | 4 | 616 | |||
XM_008769629.1 | XP_008767851.1 | 4 | 579 | |||
XM_008769630.1 | XP_008767852.1 | 4 | 579 | |||
XM_008769631.1 | XP_008767853.1 | 5 | 873 | |||
XM_008769632.1 | XP_008767854.1 | 5 | 494 | |||
GenBank mRNA | BC100650.1 | - | 6 | 952 | 7932888 |
