siRNA Details
Gene Symbol: Abl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107186.1 | NP_001100656.1 | 11, 12 | 1821 | ||
XM_006250047.2 | XP_006250109.1 | 11, 12 | 2129 | |||
XM_006250049.2 | XP_006250111.1 | 11, 12 | 2799 | |||
XM_006250051.2 | XP_006250113.1 | 10, 11 | 2066 | |||
XM_006250053.2 | XP_006250115.1 | 11, 12 | 2128 | |||
XM_006250054.2 | XP_006250116.1 | 11, 12 | 2798 | |||
XM_006250055.2 | XP_006250117.1 | 10, 11 | 2065 |
