siRNA Details
Gene Symbol: Abl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107186.1 | NP_001100656.1 | 11 | 1687 | ||
XM_006250046.2 | XP_006250108.1 | 11 | 1995 | |||
XM_006250047.2 | XP_006250109.1 | 11 | 1995 | |||
XM_006250048.2 | XP_006250110.1 | 11 | 2666 | |||
XM_006250049.2 | XP_006250111.1 | 11 | 2665 | |||
XM_006250050.2 | XP_006250112.1 | 10 | 1932 | |||
XM_006250051.2 | XP_006250113.1 | 10 | 1932 | |||
XM_006250052.2 | XP_006250114.1 | 11 | 1995 | |||
XM_006250053.2 | XP_006250115.1 | 11 | 1994 | |||
XM_006250054.2 | XP_006250116.1 | 11 | 2664 | |||
XM_006250055.2 | XP_006250117.1 | 10 | 1931 |
