siRNA Details
Gene Symbol: Tor3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009683.1 | NP_001009683.1 | 6 | 983 | ||
GenBank mRNA | BC087678.1 | - | 6 | 983 | 7318816 | |
FQ226439.1 | - | 4 | 544 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tor3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009683.1 | NP_001009683.1 | 6 | 983 | ||
GenBank mRNA | BC087678.1 | - | 6 | 983 | 7318816 | |
FQ226439.1 | - | 4 | 544 |