siRNA Details
Gene Symbol: Fam20b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107187.1 | NP_001100657.1 | 8 | 1375 | ||
XM_006250058.2 | XP_006250120.1 | 9 | 1494 | |||
XM_006250059.2 | XP_006250121.1 | 10 | 1592 | |||
XM_008769634.1 | XP_008767856.1 | 9 | 1974 |
