siRNA Details
Gene Symbol: Ralgps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100680.1 | NP_001094150.1 | 9 | 957 | ||
GenBank mRNA | BC105626.1 | - | 9 | 957 | 7326132 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_023884, NM_001159967, NM_001159968, NM_001159965, NM_001159966, and others.
Gene Ontology Categories:
