siRNA Details
Gene Symbol: Ralgps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100680.1 | NP_001094150.1 | 5 | 467 | ||
GenBank mRNA | AF296692.1 | - | 410 | |||
BC105626.1 | - | 5 | 467 | 7326132 |

More Information
Other Species Targeted

Gene Ontology Categories:
