siRNA Details
Gene Symbol: Ralgps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100680.1 | NP_001094150.1 | 8 | 861 | ||
GenBank mRNA | BC105626.1 | - | 8 | 861 | 7326132 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ralgps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100680.1 | NP_001094150.1 | 8 | 861 | ||
GenBank mRNA | BC105626.1 | - | 8 | 861 | 7326132 |