siRNA Details
Gene Symbol: Dars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034143.1 | NP_001029315.1 | 17 | 1941 | ||
GenBank mRNA | BC105774.1 | - | 17 | 1941 | 7930977 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
