siRNA Details
Gene Symbol: Tbx19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107193.1 | NP_001100663.1 | 3 | 683 | ||
XM_008769651.1 | XP_008767873.1 | 3 | 1359 | |||
XM_008769652.1 | XP_008767874.1 | 4 | 720 |

More Information
Other Species Targeted

Gene Ontology Categories:
