siRNA Details
Gene Symbol: Tbx19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107193.1 | NP_001100663.1 | 5, 6 | 1058 | ||
XM_008769651.1 | XP_008767873.1 | 6, 7 | 1796 | |||
XM_008769652.1 | XP_008767874.1 | 7, 8 | 1157 |
