siRNA Details
Gene Symbol: Pogk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107194.2 | NP_001100664.2 | 5 | 1759 | ||
XM_006250161.2 | XP_006250223.1 | 6 | 2133 | |||
XM_006250162.2 | XP_006250224.1 | 6 | 1998 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
