siRNA Details
Gene Symbol: Tagln2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013127.1 | NP_001013145.1 | 5 | 541 | ||
XM_006250295.2 | XP_006250357.1 | 5 | 976 | |||
GenBank mRNA | BC084703.1 | - | 5 | 541 | 7300611 | |
FQ221577.1 | - | 5 | 552 | |||
FQ223112.1 | - | 5 | 566 | |||
FQ228757.1 | - | 5 | 560 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
