siRNA Details
Gene Symbol: Exo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107198.1 | NP_001100668.1 | 4 | 409 | ||
XM_008769787.1 | XP_008768009.1 | 2 | 286 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Exo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107198.1 | NP_001100668.1 | 4 | 409 | ||
XM_008769787.1 | XP_008768009.1 | 2 | 286 |