siRNA Details
Gene Symbol: Sdccag8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177929.2 | NP_808790.1 | 17 | 2126 | ||
XM_006250321.2 | XP_006250383.1 | 17 | 2271 | |||
XM_006250322.2 | XP_006250384.1 | 17 | 2271 | |||
XM_008769788.1 | XP_008768010.1 | 17 | 2271 | |||
GenBank mRNA | AF352814.1 | - | 17 | 2126 | ||
BC086973.1 | - | 17 | 2122 | 7191324 |
