siRNA Details
Gene Symbol: Ptpn14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107200.1 | NP_001100670.1 | 4 | 765 | ||
XM_006250453.2 | XP_006250515.1 | 4 | 877 | |||
XM_008769853.1 | XP_008768075.1 | 3 | 495 | |||
XM_008769854.1 | XP_008768076.1 | 4 | 527 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_008976, D31842, AK086524, AK156480, AF170902, and others.
Gene Ontology Categories:
