siRNA Details
Gene Symbol: Tmem206 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007679.1 | NP_001007680.1 | 8 | 1054 | ||
XM_006250481.2 | XP_006250543.1 | 7 | 878 | |||
XM_008769855.1 | XP_008768077.1 | 8 | 1086 | |||
GenBank mRNA | BC082061.1 | - | 8 | 1054 | 7132483 | |
FQ225543.1 | - | 8 | 1063 | |||
FQ228821.1 | - | 9 | 1096 | |||
FQ233431.1 | - | 8 | 1059 |
