siRNA Details
Gene Symbol: Tmem206 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007679.1 | NP_001007680.1 | 7 | 863 | ||
XM_006250481.2 | XP_006250543.1 | 6 | 687 | |||
XM_008769855.1 | XP_008768077.1 | 7 | 895 | |||
GenBank mRNA | BC082061.1 | - | 7 | 863 | 7132483 | |
FQ225543.1 | - | 7 | 872 | |||
FQ228821.1 | - | 8 | 905 | |||
FQ233431.1 | - | 7 | 868 |
