siRNA Details
Gene Symbol: Diexf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013986.2 | NP_001014008.1 | 6 | 963 | ||
XM_006250482.2 | XP_006250544.1 | 7 | 1153 | |||
GenBank mRNA | BC087114.1 | - | 6 | 917 | 7193786 | |
FQ217506.1 | - | 7 | 1136 |
