siRNA Details
Gene Symbol: Fbxo28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107203.3 | NP_001100673.1 | 1, 2 | 284 | ||
GenBank mRNA | BC167003.1 | - | 1, 2 | 285 | 8366732 |

More Information
Other Species Targeted

