siRNA Details
Gene Symbol: Brdt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012031.2 | NP_001012031.1 | 7 | 1177 | ||
GenBank mRNA | BC078999.1 | - | 7 | 1155 | ||
BC158630.1 | - | 7 | 1152 | 7456383 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
