siRNA Details
Gene Symbol: Aff1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107206.1 | NP_001100676.1 | 16, 17 | 3247 | ||
XM_006250632.2 | XP_006250694.1 | 17, 18 | 3352 | |||
XM_006250633.2 | XP_006250695.1 | 16, 17 | 3236 |
