siRNA Details
Gene Symbol: Arhgap24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012032.1 | NP_001012032.1 | 4 | 775 | ||
XM_006250652.2 | XP_006250714.1 | 5 | 981 | |||
XM_006250654.2 | XP_006250716.1 | 5 | 3563 | |||
XM_008770017.1 | XP_008768239.1 | 5 | 1642 | |||
XM_008770018.1 | XP_008768240.1 | 4 | 1494 | |||
GenBank mRNA | AB108670.1 | - | 4 | 977 | ||
BC085797.1 | - | 4 | 775 | 7109913 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
