siRNA Details
Gene Symbol: Arhgap24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012032.1 | NP_001012032.1 | 6 | 1016 | ||
XM_006250652.2 | XP_006250714.1 | 7 | 1222 | |||
XM_006250654.2 | XP_006250716.1 | 7 | 3804 | |||
XM_008770017.1 | XP_008768239.1 | 7 | 1883 | |||
XM_008770018.1 | XP_008768240.1 | 6 | 1735 | |||
GenBank mRNA | AB108670.1 | - | 6 | 1218 | ||
BC085797.1 | - | 6 | 1016 | 7109913 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
