siRNA Details
Gene Symbol: Hnrnpdl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033696.1 | NP_001028868.1 | 4 | 680 | ||
XM_006250662.2 | XP_006250724.1 | 4 | 816 | |||
XM_006250663.2 | XP_006250725.1 | 4 | 816 | |||
XM_008770024.1 | XP_008768246.1 | 4 | 816 | |||
GenBank mRNA | BC104683.1 | - | 4 | 680 | 7461444 | |
FQ222460.1 | - | 1410 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
