siRNA Details
Gene Symbol: Thap6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107209.2 | NP_001100679.1 | 5 | 587 | ||
XM_008770033.1 | XP_008768255.1 | 4 | 528 | |||
XM_008770035.1 | XP_008768257.1 | 4 | 467 | |||
GenBank mRNA | CK602350.1 | - | 4 | 486 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
