siRNA Details
Gene Symbol: Apbb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770169.1 | XP_008768391.1 | 12 | 2038 | ||
GenBank mRNA | FQ211919.1 | - | 482 |

More Information
Other Species Targeted

Gene Ontology Categories:
