siRNA Details
Gene Symbol: Nsun7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017452.1 | NP_001017452.1 | 4 | 747 | ||
XM_006250946.2 | XP_006251008.1 | 4 | 1038 | |||
XM_006250947.2 | XP_006251009.1 | 4 | 1041 | |||
XM_006250948.2 | XP_006251010.1 | 4 | 1038 | |||
GenBank mRNA | BC083653.1 | - | 4 | 747 | 7135159 |
