siRNA Details
Gene Symbol: Rbm47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005882.1 | NP_001005882.1 | 2 | 884 | ||
XM_006250949.2 | XP_006251011.1 | 3 | 1967 | |||
XM_006250950.2 | XP_006251012.1 | 3 | 2865 | |||
XM_006250951.2 | XP_006251013.1 | 5 | 1213 | |||
XM_006250952.2 | XP_006251014.1 | 4 | 1439 | |||
XM_006250954.2 | XP_006251016.1 | 4 | 1970 | |||
XM_006250955.2 | XP_006251017.1 | 3 | 2158 | |||
XM_008770152.1 | XP_008768374.1 | 3 | 1672 | |||
GenBank mRNA | AY724525.1 | - | 3 | 907 | ||
BC081995.1 | - | 2 | 884 | 7130530 |
