siRNA Details
Gene Symbol: Pds5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083624.1 | NP_001077093.1 | 23, 24 | 2778 | ||
XM_008770153.1 | XP_008768375.1 | 23, 24 | 2812 | |||
XM_008770154.1 | XP_008768376.1 | 22, 23 | 2725 | |||
GenBank mRNA | EF460313.1 | - | 23, 24 | 2778 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
