siRNA Details
Gene Symbol: Lias | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012037.1 | NP_001012037.1 | 2 | 242 | ||
XM_008770155.1 | XP_008768377.1 | 2 | 278 | |||
XM_008770156.1 | XP_008768378.1 | 2 | 279 | |||
GenBank mRNA | BC083708.1 | - | 2 | 242 | 7190554 |
