siRNA Details
Gene Symbol: Lgi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107219.2 | NP_001100689.2 | 8 | 1480 | ||
GenBank mRNA | BC168885.1 | - | 8 | 1480 | 7121391 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lgi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107219.2 | NP_001100689.2 | 8 | 1480 | ||
GenBank mRNA | BC168885.1 | - | 8 | 1480 | 7121391 |