siRNA Details
Gene Symbol: Pi4k2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005883.1 | NP_001005883.1 | 4 | 792 | ||
GenBank mRNA | BC083668.1 | - | 4 | 792 | 7135573 |

More Information
Other Species Targeted

Gene Ontology Categories:
