siRNA Details
Gene Symbol: Pi4k2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005883.1 | NP_001005883.1 | 2 | 402 | ||
GenBank mRNA | BC083668.1 | - | 2 | 402 | 7135573 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pi4k2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005883.1 | NP_001005883.1 | 2 | 402 | ||
GenBank mRNA | BC083668.1 | - | 2 | 402 | 7135573 |