siRNA Details
Gene Symbol: Anapc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107220.1 | NP_001100690.1 | 22 | 1842 | ||
GenBank mRNA | BC086546.1 | - | 12 | 756 | ||
FQ227240.1 | - | 4 | 274 |

More Information
Other Species Targeted

Gene Ontology Categories:
