siRNA Details
Gene Symbol: Fbxl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107222.1 | NP_001100692.1 | 2 | 204 | ||
XM_006251076.2 | XP_006251138.1 | 2 | 285 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fbxl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107222.1 | NP_001100692.1 | 2 | 204 | ||
XM_006251076.2 | XP_006251138.1 | 2 | 285 |