siRNA Details
Gene Symbol: Sh3tc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107226.1 | NP_001100696.1 | 11 | 1392 | ||
XM_006251125.2 | XP_006251187.1 | 12 | 1923 | |||
XM_006251126.2 | XP_006251188.1 | 13 | 1734 | |||
XM_006251127.2 | XP_006251189.1 | 12 | 1535 | |||
XM_006251128.2 | XP_006251190.1 | 12 | 3048 | |||
XM_006251130.2 | XP_006251192.1 | 11 | 1847 | |||
XM_008770281.1 | XP_008768503.1 | 12 | 2185 | |||
XM_008770282.1 | XP_008768504.1 | 13 | 1609 |
