siRNA Details
Gene Symbol: Dusp18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013128.2 | NP_001013146.2 | 2 | 377 | ||
XM_006251299.2 | XP_006251361.1 | 2 | 801 | |||
GenBank mRNA | BC079285.1 | - | 2 | 359 | 7134264 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
