siRNA Details
Gene Symbol: Ikzf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107237.1 | NP_001100707.1 | 5 | 592 | ||
XM_006251491.2 | XP_006251553.1 | 6 | 1365 | |||
XM_006251492.2 | XP_006251554.1 | 6 | 5200 | |||
XM_006251493.2 | XP_006251555.1 | 6 | 903 | |||
XM_006251494.2 | XP_006251556.2 | 6 | 1190 | |||
XM_006251495.2 | XP_006251557.1 | 6 | 878 | |||
XM_006251496.2 | XP_006251558.1 | 6 | 677 | |||
XM_006251497.2 | XP_006251559.1 | 6 | 911 | |||
XM_008770312.1 | XP_008768534.1 | 6 | 1190 | |||
XM_008770314.1 | XP_008768536.1 | 6 | 1190 | |||
XM_008770315.1 | XP_008768537.1 | 5 | 932 | |||
XM_008770317.1 | XP_008768539.1 | 5 | 932 |
