siRNA Details
Gene Symbol: Tmed4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006251444.2 | XP_006251506.1 | 1, 2 | 209 | ||
XM_006251445.2 | XP_006251507.1 | 1, 2 | 210 | |||
GenBank mRNA | BC168726.1 | - | 1, 2 | 181 | 6889842 | |
BC168735.1 | - | 1, 2 | 181 | 6891034 | ||
BC168766.1 | - | 1, 2 | 181 | 6890852 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_134020, BC023041, AK078425, AF368921, AK158876.
Gene Ontology Categories:
