siRNA Details
Gene Symbol: B3gnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107240.1 | NP_001100710.1 | 3 | 793 | ||
XM_006251554.2 | XP_006251616.1 | 2 | 592 | |||
XM_006251555.1 | XP_006251617.1 | 2 | 476 | |||
XM_006251556.2 | XP_006251618.1 | 2 | 475 | |||
XM_008770427.1 | XP_008768649.1 | 2 | 2311 |
