siRNA Details
Gene Symbol: Ahsa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107241.1 | NP_001100711.1 | 5 | 583 | ||
XM_006251539.2 | XP_006251601.1 | 6 | 718 | |||
XM_006251540.2 | XP_006251602.1 | 5 | 606 | |||
XM_006251541.2 | XP_006251603.1 | 5 | 600 | |||
XM_006251542.2 | XP_006251604.1 | 5 | 633 | |||
GenBank mRNA | BC099114.1 | - | 6 | 622 |

More Information
Other Species Targeted

