siRNA Details
Gene Symbol: Pus10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025278.1 | NP_001020449.1 | 2, 3 | 547 | ||
XM_008770428.1 | XP_008768650.1 | 2, 3 | 491 | |||
XM_008770429.1 | XP_008768651.1 | 2, 3 | 491 | |||
GenBank mRNA | BC093611.1 | - | 2, 3 | 547 | 7307810 |
