siRNA Details
Gene Symbol: Papolg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107244.1 | NP_001100714.1 | 13, 14 | 1402 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Papolg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107244.1 | NP_001100714.1 | 13, 14 | 1402 |